Assay Details
| Gene Symbol: Mgat4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012225.2 | NP_001012225.1 | 16 | 6992 | 135 | |
| NM_001160155.1 | NP_001153627.1 | 16 | 7019 | 135 | ||
| XM_006244840.2 | XP_006244902.1 | 16 | 7029 | 135 | ||
| XM_008767040.1 | XP_008765262.1 | 15 | 6707 | 135 | ||
| GenBank mRNA | BM388934.1 | - | 1 | 135 | ||
More Information
Set Membership: |
|

Mgat4a
Genomic Map