Assay Details
| Gene Symbol: Rccd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229455.2 | XP_006229517.1 | 4 - 5 | 1406 | 66 | |
| XM_006229456.2 | XP_006229518.1 | 4 - 5 | 1062 | 66 | ||
| XM_006229457.2 | XP_006229519.1 | 4 - 5 | 1307 | 66 | ||
| XM_218819.7 | XP_218819.5 | 4 - 5 | 1307 | 66 | ||
More Information
Set Membership: |
|

Rccd1
Genomic Map