Assay Details
Gene Symbol: Nras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080766.2 | NP_542944.1 | 4 | 490 | 102 | |
XM_006233059.2 | XP_006233121.1 | 4 | 534 | 102 | ||
GenBank mRNA | BC098659.1 | - | 4 | 490 | 7368145 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
