Assay Details
Gene Symbol: LOC100362110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | S80119.1 | - | 798 | 89 |
Gene Symbol: Crp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY310153.1 | - | 824 | 89 | ||
AY318961.1 | - | 824 | 89 | |||
AY321318.1 | - | 821 | 89 | |||
AY321324.1 | - | 824 | 89 | |||
AY325196.1 | - | 824 | 89 | |||
AY325218.1 | - | 824 | 89 | |||
AY325232.1 | - | 824 | 89 |
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY387051.1 | - | 89 |
Gene Symbol: Lrrgt00014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U83119.1 | - | 1 | 2740 | 89 |
Gene Symbol: Serinc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY539942.1 | - | 2035 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
