Assay Details
| Gene Symbol: Cistr-act | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104335.1 | 1 | 176 | 60 | ||
| GenBank mRNA | BM385021.1 | - | 1 | 176 | 60 | |
| BM385392.1 | - | 1 | 176 | 60 | ||
More Information
Set Membership: |
|

Cistr-act
Genomic Map