Assay Details
Gene Symbol: Tpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053867.1 | NP_446319.1 | 6 | 623 | 93 | |
GenBank mRNA | BC086358.1 | - | 6 | 634 | 7303675 | 93 |
FQ210153.1 | - | 6 | 651 | 93 | ||
FQ210845.1 | - | 5 | 537 | 93 | ||
FQ210981.1 | - | 2 | 84 | 93 | ||
FQ217312.1 | - | 6 | 651 | 93 | ||
FQ217485.1 | - | 6 | 651 | 93 | ||
FQ220662.1 | - | 6 | 651 | 93 | ||
FQ220913.1 | - | 6 | 654 | 93 | ||
FQ221184.1 | - | 6 | 651 | 93 | ||
FQ221191.1 | - | 6 | 640 | 93 | ||
FQ221220.1 | - | 6 | 651 | 93 | ||
FQ221227.1 | - | 6 | 651 | 93 | ||
FQ221357.1 | - | 6 | 651 | 93 | ||
FQ221503.1 | - | 6 | 651 | 93 | ||
FQ221524.1 | - | 6 | 651 | 93 | ||
FQ221695.1 | - | 6 | 651 | 93 | ||
FQ221696.1 | - | 6 | 652 | 93 | ||
FQ221929.1 | - | 3 | 269 | 93 | ||
FQ222296.1 | - | 6 | 651 | 93 | ||
FQ222704.1 | - | 6 | 650 | 93 | ||
FQ222827.1 | - | 6 | 651 | 93 | ||
FQ223078.1 | - | 6 | 651 | 93 | ||
FQ223172.1 | - | 6 | 652 | 93 | ||
FQ223294.1 | - | 6 | 658 | 93 | ||
FQ223538.1 | - | 6 | 651 | 93 | ||
FQ223612.1 | - | 6 | 640 | 93 | ||
FQ224095.1 | - | 6 | 651 | 93 | ||
FQ224271.1 | - | 6 | 652 | 93 | ||
FQ224320.1 | - | 6 | 650 | 93 | ||
FQ224622.1 | - | 6 | 587 | 93 | ||
FQ228556.1 | - | 6 | 651 | 93 | ||
FQ228624.1 | - | 6 | 650 | 93 | ||
FQ228630.1 | - | 6 | 651 | 93 | ||
FQ228644.1 | - | 6 | 648 | 93 | ||
FQ229093.1 | - | 6 | 651 | 93 | ||
FQ229217.1 | - | 6 | 651 | 93 | ||
FQ229279.1 | - | 6 | 651 | 93 | ||
FQ229346.1 | - | 6 | 651 | 93 | ||
FQ229353.1 | - | 6 | 651 | 93 | ||
FQ230785.1 | - | 6 | 651 | 93 | ||
FQ231339.1 | - | 6 | 649 | 93 | ||
FQ231778.1 | - | 6 | 651 | 93 | ||
FQ232593.1 | - | 6 | 651 | 93 | ||
FQ232768.1 | - | 6 | 652 | 93 | ||
FQ233320.1 | - | 6 | 682 | 93 | ||
FQ233909.1 | - | 6 | 654 | 93 | ||
FQ234047.1 | - | 6 | 641 | 93 | ||
FQ234186.1 | - | 6 | 93 | |||
FQ234298.1 | - | 6 | 651 | 93 | ||
FQ234990.1 | - | 6 | 651 | 93 | ||
FQ235307.1 | - | 6 | 651 | 93 | ||
U20525.1 | - | 6 | 623 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
