Assay Details
Gene Symbol: Zfp52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228004.2 | XP_006228066.1 | 3 - 4 | 367 | 97 | |
XM_006228006.2 | XP_006228068.1 | 3 - 4 | 450 | 97 | ||
XM_006228007.2 | XP_006228069.1 | 2 - 3 | 2846 | 97 | ||
XM_008758764.1 | XP_008756986.1 | 3 - 4 | 207 | 97 | ||
XM_008758765.1 | XP_008756987.1 | 3 - 4 | 1798 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006228007.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
