Assay Details
| Gene Symbol: Znf761 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228010.2 | XP_006228072.1 | 1 - 2 | 1530 | 58 | |
| XM_006228011.2 | XP_006228073.1 | 1 - 2 | 1530 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006228010.2, XM_006228011.2 transcripts.
Set Membership: |
|

Znf761
Genomic Map