Assay Details
| Gene Symbol: Foxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006242968.2 | XP_006243030.1 | 2 - 3 | 558 | 84 | |
| XM_006242969.2 | XP_006243031.1 | 2 - 3 | 558 | 84 | ||
More Information
Set Membership: |
|

Foxr1
Genomic Map