Assay Details
| Gene Symbol: Dctpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138892.1 | NP_620247.1 | 3 | 546 | 92 | |
| GenBank mRNA | AF331839.1 | - | 3 | 520 | 92 | |
| AY029335.1 | - | 3 | 546 | 92 | ||
More Information
Set Membership: |
|

Dctpp1
Genomic Map