Assay Details
| Gene Symbol: Ugt1a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201425.2 | NP_958828.1 | 1 | 217 | 68 | |
| GenBank mRNA | AY435129.1 | - | 1 | 217 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Ugt1a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201425.2 | NP_958828.1 | 1 | 217 | 68 | |
| GenBank mRNA | AY435129.1 | - | 1 | 217 | 68 | |
Set Membership: |
|