Assay Details
| Gene Symbol: Ehd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011939.2 | NP_001011939.1 | 5 | 1858 | 70 | |
| GenBank mRNA | BC082030.1 | - | 5 | 1812 | 70 | |
| BC160908.1 | - | 5 | 1859 | 70 | ||
More Information
Set Membership: |
|

Ehd1
Genomic Map