Assay Details
Gene Symbol: Abi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173143.1 | NP_775166.1 | 8 | 1775 | 100 | |
GenBank mRNA | FQ222350.1 | - | 2133 | 100 | ||
U94904.1 | - | 8 | 1775 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
