Assay Details
Gene Symbol: LOC103694558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773697.1 | XP_008771919.1 | 2542 | 109 | ||
XM_008773698.1 | XP_008771920.1 | 2542 | 109 | |||
XM_008773699.1 | XP_008771921.1 | 2407 | 109 | |||
XM_008773700.1 | XP_008771922.1 | 2407 | 109 |
Gene Symbol: Kdm6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FJ775728.1 | - | 3316 | 109 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
