Assay Details
| Gene Symbol: Zfp444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228186.2 | XP_006228248.1 | 5 | 2050 | 85 | |
| XM_008758902.1 | XP_008757124.1 | 5 | 2070 | 85 | ||
| XM_008758903.1 | XP_008757125.1 | 4 | 1555 | 85 | ||
More Information
Set Membership: |
|

Zfp444
Genomic Map