Assay Details
| Gene Symbol: Gjc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100784.1 | NP_001094254.1 | 2 | 1446 | 87 | |
| XM_006246512.2 | XP_006246574.1 | 3 | 2231 | 87 | ||
| XM_006246513.2 | XP_006246575.1 | 3 | 1622 | 87 | ||
| XM_006246515.2 | XP_006246577.1 | 2 | 2028 | 87 | ||
| XM_008767849.1 | XP_008766071.1 | 3 | 1867 | 87 | ||
More Information
Set Membership: |
|

Gjc2
Genomic Map