Assay Details
Gene Symbol: Cd300c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002727843.4 | XP_002727889.2 | 2 - 3 | 600 | 101 |
Gene Symbol: RGD1565046 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768425.1 | XP_008766647.1 | 2 - 3 | 600 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
