Assay Details
| Gene Symbol: Trim80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768412.1 | XP_008766634.1 | 2 - 3 | 2059 | 71 | |
| GenBank mRNA | BC091694.1 | - | 1 - 2 | 33 | 71 | |
More Information
Set Membership: |
|

Trim80
Genomic Map