Assay Details
| Gene Symbol: Trim80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768412.1 | XP_008766634.1 | 4 - 5 | 2771 | 66 | |
| GenBank mRNA | BC091694.1 | - | 3 - 4 | 745 | 66 | |
More Information
Set Membership: |
|

Trim80
Genomic Map