Assay Details
Gene Symbol: LOC100911529 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769019.1 | XP_008767241.1 | 2 - 3 | 220 | 91 |
Gene Symbol: LOC100363914 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769020.1 | XP_008767242.1 | 3 - 4 | 220 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
