Assay Details
| Gene Symbol: Zfp316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768963.1 | XP_008767185.1 | 1 | 1427 | 112 | |
| XM_008768964.1 | XP_008767186.1 | 1 | 1427 | 112 | ||
| XM_008768965.1 | XP_008767187.1 | 1 | 1427 | 112 | ||
| XM_008768966.1 | XP_008767188.1 | 1 | 1427 | 112 | ||
| XM_008768967.1 | XP_008767189.1 | 1 | 1427 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008768963.1, XM_008768964.1, XM_008768965.1, XM_008768966.1, XM_008768967.1 transcripts.
Set Membership: |
|

Zfp316
Genomic Map