Assay Details
| Gene Symbol: Jund | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286966.1 | NP_001273895.1 | 1 | 908 | 72 | |
| NM_138875.4 | NP_620230.1 | 1 | 908 | 72 | ||
| GenBank mRNA | BC062053.1 | - | 1 | 908 | 5598945 | 72 |
More Information
Set Membership: |
|

Jund
Genomic Map