Assay Details
| Gene Symbol: Zfp646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759939.1 | XP_008758161.1 | 1 | 1246 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008759939.1 transcripts.
Set Membership: |
|

Zfp646
Genomic Map