Assay Details
| Gene Symbol: LOC690182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759848.1 | XP_008758070.1 | 1 | 1220 | 77 | |
| Gene Symbol: Egln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772679.1 | XP_008770901.1 | 7 | 2191 | 77 | |
| GenBank mRNA | BC081694.1 | - | 5 | 1220 | 77 | |
More Information
Set Membership: |
|

LOC690182
Genomic Map