Assay Details
| Gene Symbol: Scgb2b24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_577783.2 | XP_577783.2 | 1 - 2 | 58 | 86 | |
| GenBank mRNA | GU269242.1 | - | 1 - 2 | 58 | 86 | |
More Information
Set Membership: |
|

Scgb2b24
Genomic Map