Assay Details
| Gene Symbol: Serpinb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031642.1 | NP_001026812.1 | 6 | 1076 | 88 | |
| GenBank mRNA | BC098686.1 | - | 6 | 1076 | 7385870 | 88 |
More Information
Set Membership: |
|

Serpinb1a
Genomic Map