Assay Details
Gene Symbol: Ndufa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047862.1 | NP_001041327.1 | 4 | 377 | 98 | |
XM_008761741.1 | XP_008759963.1 | 3 | 405 | 98 | ||
GenBank mRNA | AY325167.1 | - | 4 | 377 | 98 | |
FQ226210.1 | - | 2 | 239 | 98 | ||
FQ230936.1 | - | 2 | 239 | 98 | ||
FQ234165.1 | - | 2 | 239 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
