Assay Details
| Gene Symbol: Zfp426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079943.1 | NP_001073412.1 | 8 | 1993 | 124 | |
| XM_006242666.1 | XP_006242728.1 | 7 | 2000 | 124 | ||
| XM_006242667.2 | XP_006242729.1 | 7 | 4088 | 124 | ||
| XM_006242668.2 | XP_006242730.1 | 8 | 3394 | 124 | ||
| XM_006242670.1 | XP_006242732.1 | 8 | 2083 | 124 | ||
| XM_006242671.2 | XP_006242733.1 | 6 | 1869 | 124 | ||
| XM_006242672.2 | XP_006242734.1 | 7 | 3596 | 124 | ||
| XM_006242673.2 | XP_006242735.1 | 6 | 3449 | 124 | ||
| XM_008765981.1 | XP_008764203.1 | 7 | 3599 | 124 | ||
| XM_008765982.1 | XP_008764204.1 | 7 | 1936 | 124 | ||
| XM_008765983.1 | XP_008764205.1 | 7 | 3244 | 124 | ||
| XM_008765984.1 | XP_008764206.1 | 7 | 3200 | 124 | ||
| GenBank mRNA | BC129125.1 | - | 8 | 1993 | 7133611 | 124 |
More Information
Set Membership: |
|

Zfp426
Genomic Map