Assay Details
| Gene Symbol: Ngfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012610.2 | NP_036742.2 | 1 | 65 | 51 | |
| GenBank mRNA | CB725789.1 | - | 1 | 73 | 51 | |
| X05137.1 | - | 1 | 65 | 51 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_012610.2, X05137.1 transcripts.
Set Membership: |
|

Ngfr
Genomic Map