Assay Details
| Gene Symbol: Pld6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246540.2 | XP_006246602.1 | 1 | 270 | 104 | |
| XM_220517.6 | XP_220517.1 | 1 | 266 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006246540.2, XM_220517.6 transcripts.
Set Membership: |
|

Pld6
Genomic Map