Assay Details
| Gene Symbol: Nsa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134415.2 | NP_599242.2 | 5 | 743 | 104 | |
| GenBank mRNA | BC098657.1 | - | 5 | 734 | 7378282 | 104 |
| BP485557.1 | - | 2 | 104 | |||
| BP486932.1 | - | 2 | 238 | 104 | ||
| DQ354588.1 | - | 5 | 733 | 104 | ||
| FQ212851.1 | - | 5 | 755 | 104 | ||
| FQ214845.1 | - | 5 | 761 | 104 | ||
| FQ225571.1 | - | 5 | 756 | 104 | ||
| FQ226635.1 | - | 5 | 770 | 104 | ||
| FQ229542.1 | - | 5 | 761 | 104 | ||
| Y17325.1 | - | 5 | 715 | 104 | ||
More Information
Set Membership: |
|

Nsa2
Genomic Map