Assay Details
Gene Symbol: Gpatch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011909.1 | NP_001011909.1 | 5 - 6 | 1210 | 65 | |
XM_006250435.2 | XP_006250497.1 | 4 - 5 | 1092 | 65 | ||
XM_006250439.2 | XP_006250501.1 | 4 - 5 | 1141 | 65 | ||
XM_008769838.1 | XP_008768060.1 | 5 - 6 | 1209 | 65 | ||
XM_008769839.1 | XP_008768061.1 | 5 - 6 | 3859 | 65 | ||
XM_008769840.1 | XP_008768062.1 | 5 - 6 | 1294 | 65 | ||
XM_008769841.1 | XP_008768063.1 | 6 - 7 | 1400 | 65 | ||
XM_008769842.1 | XP_008768064.1 | 5 - 6 | 1209 | 65 | ||
XM_008769843.1 | XP_008768065.1 | 5 - 6 | 1209 | 65 | ||
XM_008769844.1 | XP_008768066.1 | 5 - 6 | 1209 | 65 | ||
XM_008769845.1 | XP_008768067.1 | 5 - 6 | 1209 | 65 | ||
XM_008769846.1 | XP_008768068.1 | 4 - 5 | 1280 | 65 | ||
GenBank mRNA | BC079232.1 | - | 5 - 6 | 1210 | 7132530 | 65 |
FQ230498.1 | - | 4 - 5 | 1141 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
