Assay Details
| Gene Symbol: Xkr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012092.1 | NP_001012092.1 | 2 - 3 | 951 | 82 | |
| XM_008762334.1 | XP_008760556.1 | 3 - 4 | 2675 | 82 | ||
| GenBank mRNA | AY534261.1 | - | 2 - 3 | 951 | 82 | |
More Information
Set Membership: |
|

Xkr7
Genomic Map