Assay Details
| Gene Symbol: Cmtm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013142.1 | NP_001013160.1 | 1 - 2 | 268 | 78 | |
| GenBank mRNA | BC078975.1 | - | 1 - 2 | 268 | 7111936 | 78 |
More Information
Set Membership: |
|

Cmtm2a
Genomic Map