Assay Details
| Gene Symbol: Apol3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013175.1 | NP_001013193.1 | 2 - 3 | 134 | 75 | |
| NM_001277359.1 | NP_001264288.1 | 2 - 3 | 216 | 75 | ||
| GenBank mRNA | BC086447.1 | - | 2 - 3 | 134 | 7304352 | 75 |
| FQ231474.1 | - | 2 - 3 | 232 | 75 | ||
| FQ233422.1 | - | 2 - 3 | 298 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC086447.1, NM_001013175.1, NM_001277359.1 transcripts.
Set Membership: |
|

Apol3
Genomic Map