Assay Details
Gene Symbol: Tmem171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013903.1 | NP_001013925.1 | 2 - 3 | 742 | 95 | |
XM_006231833.2 | XP_006231895.1 | 2 - 3 | 1659 | 95 | ||
XM_006231834.2 | XP_006231896.1 | 2 - 3 | 1658 | 95 | ||
XM_008760660.1 | XP_008758882.1 | 2 - 3 | 1175 | 95 | ||
GenBank mRNA | AY185507.1 | - | 2 - 3 | 742 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
