Assay Details
| Gene Symbol: Cyb561a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014164.1 | NP_001014186.1 | 2 - 3 | 190 | 62 | |
| XM_006231049.2 | XP_006231111.1 | 2 - 3 | 790 | 62 | ||
| XM_006231050.2 | XP_006231112.1 | 2 - 3 | 658 | 62 | ||
| XM_006231051.2 | XP_006231113.1 | 2 - 3 | 623 | 62 | ||
| XM_006231052.2 | XP_006231114.1 | 2 - 3 | 825 | 62 | ||
| XM_006231053.2 | XP_006231115.1 | 2 - 3 | 209 | 62 | ||
| XM_006231054.2 | XP_006231116.1 | 2 - 3 | 169 | 62 | ||
| XM_008760233.1 | XP_008758455.1 | 2 - 3 | 545 | 62 | ||
| XM_008760234.1 | XP_008758456.1 | 2 - 3 | 153 | 62 | ||
| GenBank mRNA | BC085835.1 | - | 2 - 3 | 190 | 7134938 | 62 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC085835.1, NM_001014164.1, XM_006231050.2, XM_006231051.2, XM_006231052.2, XM_006231053.2, XM_006231054.2, XM_008760233.1, XM_008760234.1 transcripts.
Set Membership: |
|

Cyb561a3
Genomic Map