Assay Details
| Gene Symbol: Cyb561a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014164.1 | NP_001014186.1 | 3 - 4 | 397 | 51 | |
| XM_006231049.2 | XP_006231111.1 | 3 - 4 | 997 | 51 | ||
| XM_006231050.2 | XP_006231112.1 | 3 - 4 | 865 | 51 | ||
| XM_006231051.2 | XP_006231113.1 | 3 - 4 | 830 | 51 | ||
| XM_006231052.2 | XP_006231114.1 | 3 - 4 | 1032 | 51 | ||
| XM_006231053.2 | XP_006231115.1 | 3 - 4 | 416 | 51 | ||
| XM_006231054.2 | XP_006231116.1 | 3 - 4 | 376 | 51 | ||
| XM_006231055.2 | XP_006231117.1 | 2 - 3 | 579 | 51 | ||
| XM_008760233.1 | XP_008758455.1 | 3 - 4 | 752 | 51 | ||
| XM_008760234.1 | XP_008758456.1 | 3 - 4 | 360 | 51 | ||
| GenBank mRNA | BC085835.1 | - | 3 - 4 | 397 | 7134938 | 51 |
More Information
Set Membership: |
|

Cyb561a3
Genomic Map