Assay Details
| Gene Symbol: Cyb561a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014164.1 | NP_001014186.1 | 5 - 6 | 760 | 94 | |
| XM_006231049.2 | XP_006231111.1 | 5 - 6 | 1360 | 94 | ||
| XM_006231050.2 | XP_006231112.1 | 5 - 6 | 1228 | 94 | ||
| XM_006231051.2 | XP_006231113.1 | 5 - 6 | 1193 | 94 | ||
| XM_006231052.2 | XP_006231114.1 | 5 - 6 | 1395 | 94 | ||
| XM_006231053.2 | XP_006231115.1 | 5 - 6 | 779 | 94 | ||
| XM_006231054.2 | XP_006231116.1 | 5 - 6 | 739 | 94 | ||
| XM_006231055.2 | XP_006231117.1 | 4 - 5 | 942 | 94 | ||
| XM_008760233.1 | XP_008758455.1 | 5 - 6 | 1115 | 94 | ||
| XM_008760234.1 | XP_008758456.1 | 5 - 6 | 723 | 94 | ||
| GenBank mRNA | BC085835.1 | - | 5 - 6 | 760 | 7134938 | 94 |
More Information
Set Membership: |
|

Cyb561a3
Genomic Map