Assay Details
| Gene Symbol: Cxcl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017496.1 | NP_001017496.1 | 2 - 3 | 218 | 70 | |
| XM_006250710.2 | XP_006250772.1 | 2 - 3 | 256 | 70 | ||
| GenBank mRNA | BC088260.1 | - | 2 - 3 | 218 | 7375992 | 70 |
More Information
Set Membership: |
|

Cxcl13
Genomic Map