Assay Details
| Gene Symbol: Il2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013163.1 | NP_037295.1 | 5 - 6 | 759 | 108 | |
| GenBank mRNA | FQ228242.1 | - | 791 | 108 | ||
| M55049.1 | - | 5 - 6 | 759 | 108 | ||
More Information
Set Membership: |
|

Il2ra
Genomic Map