Assay Details
| Gene Symbol: Pgrmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021766.1 | NP_068534.1 | 2 - 3 | 588 | 137 | |
| GenBank mRNA | AF163321.1 | - | 2 - 3 | 546 | 132 | |
| AJ005837.1 | - | 2 - 3 | 560 | 132 | ||
| BC062073.1 | - | 2 - 3 | 549 | 5599449 | 132 | |
| U63315.1 | - | 2 - 3 | 588 | 137 | ||
More Information
Set Membership: |
|

Pgrmc1
Genomic Map