Assay Details
| Gene Symbol: Npffr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023980.1 | NP_076470.1 | 1 - 2 | 326 | 62 | |
| XM_006250787.2 | XP_006250849.1 | 2 - 3 | 887 | 62 | ||
| GenBank mRNA | AF268900.1 | - | 1 - 2 | 326 | 62 | |
More Information
Set Membership: |
|

Npffr2
Genomic Map