Assay Details
| Gene Symbol: Pdgfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031317.1 | NP_112607.1 | 5 - 6 | 924 | 84 | |
| GenBank mRNA | AB033830.1 | - | 5 - 6 | 924 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Pdgfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031317.1 | NP_112607.1 | 5 - 6 | 924 | 84 | |
| GenBank mRNA | AB033830.1 | - | 5 - 6 | 924 | 84 | |
Set Membership: |
|