Assay Details
| Gene Symbol: Ramp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031646.1 | NP_113834.1 | 1 - 2 | 140 | 89 | |
| GenBank mRNA | AB030943.1 | - | 1 - 2 | 149 | 89 | |
| AB042888.1 | - | 1 - 2 | 140 | 89 | ||
| AF181551.1 | - | 1 - 2 | 143 | 89 | ||
| FQ226087.1 | - | 1 - 2 | 165 | 89 | ||
More Information
Set Membership: |
|

Ramp2
Genomic Map