Assay Details
| Gene Symbol: Uba52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031687.3 | NP_113875.1 | 1 - 2 | 39 | 101 | |
| GenBank mRNA | BP480615.1 | - | 3 - 4 | 445 | 101 | |
| FQ217153.1 | - | 1 - 2 | 40 | 101 | ||
| FQ222440.1 | - | 1 - 2 | 40 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_031687.3 transcripts.
Set Membership: |
|

Uba52
Genomic Map