Assay Details
| Gene Symbol: Ptpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053990.1 | NP_446442.1 | 4 - 5 | 380 | 66 | |
| XM_006254854.2 | XP_006254916.1 | 4 - 5 | 1619 | 66 | ||
| XM_006254855.2 | XP_006254917.1 | 4 - 5 | 1619 | 66 | ||
| XM_008772080.1 | XP_008770302.1 | 4 - 5 | 1619 | 66 | ||
| XM_008772081.1 | XP_008770303.1 | 4 - 5 | 1619 | 66 | ||
| GenBank mRNA | BC078758.1 | - | 4 - 5 | 419 | 7132485 | 66 |
| X58828.1 | - | 4 - 5 | 380 | 66 | ||
More Information
Set Membership: |
|

Ptpn2
Genomic Map