Assay Details
Gene Symbol: Csrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057144.1 | NP_476485.1 | 2 - 3 | 321 | 54 | |
XM_006229237.2 | XP_006229299.1 | 3 - 4 | 425 | 54 | ||
GenBank mRNA | X81193.1 | - | 2 - 3 | 321 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
