Assay Details
| Gene Symbol: Chrnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133597.1 | NP_598281.1 | 4 - 5 | 556 | 113 | |
| GenBank mRNA | AY574259.1 | - | 4 - 5 | 379 | 113 | |
| J04636.1 | - | 4 - 5 | 556 | 113 | ||
More Information
Set Membership: |
|

Chrnb3
Genomic Map