Assay Details
| Gene Symbol: Chrnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133597.1 | NP_598281.1 | 5 - 6 | 1436 | 87 | |
| GenBank mRNA | AY574259.1 | - | 5 - 6 | 1259 | 87 | |
| J04636.1 | - | 5 - 6 | 1436 | 87 | ||
More Information
Set Membership: |
|

Chrnb3
Genomic Map